Assay Details
| Gene Symbol: Copb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015827.2 | NP_056642.1 | 18 - 19 | 2389 | 85 | |
| GenBank mRNA | AF043120.1 | - | 7 - 8 | 934 | 85 | |
| AK088064.1 | - | 18 - 19 | 2390 | 85 | ||
| AK153352.1 | - | 18 - 19 | 2390 | 85 | ||
| AK205065.1 | - | 2 - 3 | 122 | 85 | ||
| BC006675.1 | - | 18 - 19 | 2376 | 3499585 | 85 | |
| BC010842.1 | - | 18 - 19 | 2378 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Copb2
Genomic Map