Assay Details
Gene Symbol: Npepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213733.2 | NP_998898.1 | 10 - 11 | 1374 | 87 | |
GenBank mRNA | AK129470.1 | - | 1 - 2 | 563 | 87 | |
AK150313.1 | - | 10 - 11 | 1375 | 87 | ||
AK156136.1 | - | 10 - 11 | 1375 | 87 | ||
AK170509.1 | - | 10 - 11 | 1375 | 87 | ||
BC023239.1 | - | 9 - 10 | 1052 | 87 | ||
BC055448.1 | - | 8 - 9 | 1328 | 87 | ||
BC069933.1 | - | 10 - 11 | 1369 | 4008091 | 87 | |
BC080707.1 | - | 10 - 11 | 1247 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
