Assay Details
| Gene Symbol: Npepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213733.2 | NP_998898.1 | 3 - 4 | 578 | 60 | |
| GenBank mRNA | AK144651.1 | - | 3 - 4 | 551 | 60 | |
| AK150313.1 | - | 3 - 4 | 579 | 60 | ||
| AK156136.1 | - | 3 - 4 | 579 | 60 | ||
| AK170509.1 | - | 3 - 4 | 579 | 60 | ||
| AK210473.1 | - | 1 - 2 | 83 | 60 | ||
| BC023239.1 | - | 2 - 3 | 256 | 60 | ||
| BC055448.1 | - | 2 - 3 | 253 | 60 | ||
| BC069933.1 | - | 3 - 4 | 573 | 4008091 | 60 | |
| BC080707.1 | - | 3 - 4 | 451 | 60 | ||
| BU148687.1 | - | 3 - 4 | 580 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npepl1
Genomic Map