Assay Details
| Gene Symbol: Npepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213733.2 | NP_998898.1 | 7 - 8 | 974 | 67 | |
| GenBank mRNA | AK150313.1 | - | 7 - 8 | 975 | 67 | |
| AK156136.1 | - | 7 - 8 | 975 | 67 | ||
| AK170509.1 | - | 7 - 8 | 975 | 67 | ||
| BC023239.1 | - | 6 - 7 | 652 | 67 | ||
| BC055448.1 | - | 6 - 7 | 649 | 67 | ||
| BC069933.1 | - | 7 - 8 | 969 | 4008091 | 67 | |
| BC080707.1 | - | 7 - 8 | 847 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npepl1
Genomic Map