Assay Details
| Gene Symbol: Npepl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213733.2 | NP_998898.1 | 9 - 10 | 1198 | 60 | |
| GenBank mRNA | AK150313.1 | - | 9 - 10 | 1199 | 60 | |
| AK170509.1 | - | 9 - 10 | 1199 | 60 | ||
| AK184203.1 | - | 2 - 3 | 156 | 60 | ||
| AK202709.1 | - | 2 - 3 | 156 | 60 | ||
| BC023239.1 | - | 8 - 9 | 876 | 60 | ||
| BC069933.1 | - | 9 - 10 | 1193 | 4008091 | 60 | |
| BC080707.1 | - | 9 - 10 | 1071 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Npepl1
Genomic Map