Assay Details
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022325.4 | NP_071720.1 | 3 - 4 | 594 | 58 | |
GenBank mRNA | AF136277.1 | - | 3 - 4 | 604 | 58 | |
AF197479.1 | - | 3 - 4 | 521 | 58 | ||
AJ242663.1 | - | 3 - 4 | 604 | 58 | ||
AK002710.1 | - | 3 - 4 | 556 | 58 | ||
AK004095.1 | - | 3 - 4 | 592 | 58 | ||
AK008370.1 | - | 3 - 4 | 523 | 58 | ||
AK010912.1 | - | 3 - 4 | 590 | 58 | ||
AK150329.1 | - | 3 - 4 | 522 | 58 | ||
AK150894.1 | - | 3 - 4 | 529 | 58 | ||
AK150947.1 | - | 3 - 4 | 583 | 58 | ||
AK151440.1 | - | 3 - 4 | 533 | 58 | ||
AK151585.1 | - | 3 - 4 | 557 | 58 | ||
AK151994.1 | - | 3 - 4 | 567 | 58 | ||
AK153147.1 | - | 3 - 4 | 529 | 58 | ||
AK159118.1 | - | 3 - 4 | 540 | 58 | ||
AK159618.1 | - | 3 - 4 | 607 | 58 | ||
AK159826.1 | - | 3 - 4 | 529 | 58 | ||
AK160001.1 | - | 3 - 4 | 577 | 58 | ||
AK160028.1 | - | 3 - 4 | 577 | 58 | ||
AK160354.1 | - | 3 - 4 | 609 | 58 | ||
AK166047.1 | - | 3 - 4 | 578 | 58 | ||
AK170963.1 | - | 3 - 4 | 577 | 58 | ||
BC008619.1 | - | 3 - 4 | 594 | 3588457 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
