Assay Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081338.1 | NP_001074807.1 | 13 - 14 | 1642 | 92 | |
| GenBank mRNA | AK049859.1 | - | 13 - 14 | 1697 | 92 | |
| AK220350.1 | - | 11 - 12 | 1390 | 92 | ||
| BC116639.1 | - | 12 - 13 | 1601 | 40061282 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl1
Genomic Map