Assay Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081338.1 | NP_001074807.1 | 16 - 17 | 1917 | 100 | |
| GenBank mRNA | AK049859.1 | - | 16 - 17 | 1972 | 100 | |
| AK220350.1 | - | 14 - 15 | 1665 | 100 | ||
| BC116639.1 | - | 15 - 16 | 1876 | 40061282 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl1
Genomic Map