Assay Details
| Gene Symbol: L3mbtl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081338.1 | NP_001074807.1 | 20 - 21 | 2380 | 76 | |
| GenBank mRNA | AK049859.1 | - | 19 - 20 | 2759 | 76 | |
| AK220350.1 | - | 18 - 19 | 2128 | 76 | ||
| BC116639.1 | - | 17 - 18 | 2830 | 40061282 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

L3mbtl1
Genomic Map