Assay Details
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007658.3 | NP_031684.3 | 12 - 13 | 1574 | 76 | |
GenBank mRNA | AK036499.1 | - | 2 - 3 | 1026 | 76 | |
AK077609.1 | - | 12 - 13 | 1192 | 76 | ||
AK077991.1 | - | 12 - 13 | 1576 | 76 | ||
AK142334.1 | - | 11 - 12 | 1849 | 76 | ||
BC046296.1 | - | 12 - 13 | 1193 | 6486343 | 76 | |
U27323.1 | - | 12 - 13 | 1163 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
