Assay Details
Gene Symbol: Cdc25a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007658.3 | NP_031684.3 | 7 - 8 | 1066 | 76 | |
GenBank mRNA | AK077609.1 | - | 7 - 8 | 683 | 76 | |
AK077991.1 | - | 7 - 8 | 1069 | 76 | ||
AK142334.1 | - | 6 - 7 | 1341 | 76 | ||
AK213270.1 | - | 3 - 4 | 278 | 76 | ||
BC046296.1 | - | 7 - 8 | 685 | 6486343 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
