Assay Details
| Gene Symbol: Nme6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018757.1 | NP_061227.1 | 1 - 2 | 113 | 66 | |
| GenBank mRNA | AF051942.1 | - | 1 - 2 | 113 | 66 | |
| BC002007.1 | - | 1 - 2 | 113 | 3593804 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF051942.1, BC002007.1, NM_018757.1 transcripts.
Set Membership: |
|

Nme6
Genomic Map