Assay Details
| Gene Symbol: Clec3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007223.3 | NP_001007224.1 | 2 - 3 | 255 | 99 | |
| GenBank mRNA | BC147139.1 | - | 2 - 3 | 255 | 9056169 | 99 |
| BC147140.1 | - | 2 - 3 | 255 | 9056166 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155751 | BC147140.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155751 | NM_001007223.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2155751 | BC147139.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872314 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150961 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 171439 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P168285 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155751 | BC147140.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155751 | NM_001007223.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2155751 | BC147139.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184117 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184108 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184133 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184135 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184111 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184131 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184110 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26184107 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P168285 | - | - |
| Whole Mouse Genome G4122A | A_52_P366005 | BC147140.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366005 | NM_001007223.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P366005 | BC147139.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P168285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07368561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07368554 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07361697 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07368558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26184163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184151 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07308145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184150 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184091 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184097 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06706018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07184644 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07185435 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06800430 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184106 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06760413 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07442110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184167 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06719542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184102 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184088 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184112 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07275780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06800434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184116 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184122 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184090 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184087 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184085 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184152 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184162 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07368553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184098 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184153 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07185438 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184092 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26184166 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575704 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575705 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575702 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011196.mm.1 | NM_001007223.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011196.mm.1 | NM_001007223.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011196.mm.1 | TC0800001361.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011198.mm.1 | NM_001007223.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011198.mm.1 | NM_001007223.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011198.mm.1 | TC0800001361.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2533317 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2922596 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2922596 | - | - |

Clec3a
Genomic Map