Assay Details
Gene Symbol: Sun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006225.1 | - | 10 - 11 | 883 | 75 | |
AY307077.1 | - | 10 - 11 | 902 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
