Assay Details
Gene Symbol: Bpifb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025631.3 | NP_079907.2 | 4 - 5 | 425 | 59 | |
GenBank mRNA | AK009346.1 | - | 4 - 5 | 398 | 59 | |
AK009504.1 | - | 4 - 5 | 400 | 59 | ||
AK009619.1 | - | 4 - 5 | 398 | 59 | ||
AK009776.1 | - | 4 - 5 | 382 | 59 | ||
AK009794.1 | - | 4 - 5 | 396 | 59 | ||
AK009871.1 | - | 4 - 5 | 400 | 59 | ||
AK010130.1 | - | 4 - 5 | 382 | 59 | ||
AK010154.1 | - | 4 - 5 | 396 | 59 | ||
AK010157.1 | - | 4 - 5 | 426 | 59 | ||
AK010215.1 | - | 4 - 5 | 396 | 59 | ||
AK028158.1 | - | 4 - 5 | 399 | 59 | ||
AK075874.1 | - | 4 - 5 | 396 | 59 | ||
AK160516.1 | - | 4 - 5 | 396 | 59 | ||
BC116681.1 | - | 4 - 5 | 380 | 40106489 | 59 | |
BC116727.1 | - | 4 - 5 | 380 | 40106498 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
