Assay Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080463.3 | NP_536711.3 | 5 - 6 | 808 | 109 | |
GenBank mRNA | AF375885.1 | - | 5 - 6 | 777 | 109 | |
AK044629.1 | - | 5 - 6 | 786 | 109 | ||
AK081059.1 | - | 5 - 6 | 765 | 109 | ||
AK132301.1 | - | 5 - 6 | 763 | 109 | ||
AK135635.1 | - | 5 - 6 | 790 | 109 | ||
AK170038.1 | - | 5 - 6 | 763 | 109 | ||
AK220326.1 | - | 1 - 2 | 2233 | 109 | ||
BC046295.1 | - | 5 - 6 | 808 | 6485896 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
