Assay Details
Gene Symbol: Pofut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080463.3 | NP_536711.3 | 6 - 7 | 1050 | 74 | |
GenBank mRNA | AF375885.1 | - | 6 - 7 | 1019 | 74 | |
AK044629.1 | - | 6 - 7 | 1028 | 74 | ||
AK053648.1 | - | 2 - 3 | 2553 | 74 | ||
AK081059.1 | - | 6 - 7 | 1007 | 74 | ||
AK132301.1 | - | 6 - 7 | 1005 | 74 | ||
AK220326.1 | - | 2 - 3 | 2475 | 74 | ||
BC046295.1 | - | 6 - 7 | 1050 | 6485896 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
somitogenesis
carbohydrate metabolic process
fucose metabolic process
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
fucosylation
protein O-linked fucosylation
fucosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
peptide-O-fucosyltransferase activity
angiogenesis
somitogenesis
carbohydrate metabolic process
fucose metabolic process
protein O-linked glycosylation
Notch signaling pathway
nervous system development
heart development
fucosylation
protein O-linked fucosylation
fucosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
peptide-O-fucosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753790 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753791 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753793 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106115 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591414 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 612722 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 280133 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 668901 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843529 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858509 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215826 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 4801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972714 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115740 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 372954 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 599945 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 732499 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587106 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568113 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 68910 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 828412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 868714 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13333 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 69710 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 106864 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P118409 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P176411 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154570 | BC046295.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154570 | AK081059.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154570 | AK132301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154570 | AK135635.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154570 | AK170038.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996299 | CB521087.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996299 | AK132301.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996299 | NM_080463.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753806 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753816 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753800 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753815 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753805 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753814 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753790 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753802 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753807 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753791 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753808 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753799 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753811 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753813 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753796 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21753794 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P118409 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00529942 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P176411 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00529952 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00529937 | - | - |
Whole Mouse Genome G4122A | A_52_P665833 | AK081059.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665833 | AK132301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665833 | AK220326.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665833 | AK044629.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P665833 | AK053648.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P183432 | AK081059.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P183432 | AK132301.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P183432 | NM_080463.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112108 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P111705 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P176411 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P118409 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01645620 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01563207 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753793 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753790 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21753791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753913 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753877 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753910 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753960 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21753925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753816 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01676772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753800 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753795 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01645620 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01563207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01273241 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02045754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02045758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01162395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21753804 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10477287 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477290 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477288 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477296 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477292 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477293 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477295 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477294 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10477289 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1443881_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1450137_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1456323_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10477286 | - | - |
Mouse Genome 430 2.0 Array | 1450137_at | - | - |
Mouse Genome 430 2.0 Array | 1456323_at | - | - |
Mouse Genome 430 2.0 Array | 1443881_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1456323_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1443881_at | - | - |
Mouse Genome 430 2.0 Array | 1450137_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 140484_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 140484_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200018175.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018175.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018175.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018177.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018177.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018177.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018183.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018183.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018183.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018178.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018178.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018178.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018179.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018179.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018179.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018182.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018182.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018182.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018180.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018180.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018180.mm.1 | TC0200002312.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200018181.mm.1 | NM_080463.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018181.mm.1 | NM_080463.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200018181.mm.1 | TC0200002312.mm.1 | MTA TC |
Mouse Expression Set 430 | 1456323_PM_at | - | - |
Mouse Expression Set 430 | 1450137_PM_at | - | - |
Mouse Expression Set 430 | 1450137_at | - | - |
Mouse Expression Set 430 | 1456323_at | - | - |
Mouse Expression Set 430 | 1443881_at | - | - |
Mouse Expression Set 430 | 1443881_PM_at | - | - |
Murine Genome U74v2 Set | 133811_PM_at | - | - |
Murine Genome U74v2 Set | 133811_at | - | - |
Mouse Genome 430A 2.0 Array | 1456323_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1456323_at | - | - |
Mouse Genome 430A 2.0 Array | 1450137_at | - | - |
Mouse Genome 430A 2.0 Array | 1450137_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450137_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456323_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1450137_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1456323_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443881_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1443881_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 638544 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1570458 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1570458 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001570458 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599360 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2599360 | - | - |
mouse-6 v1.1 (Illumina) | scl00140484.1_147-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1570458 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl00140484.1_147-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1570458 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3132204 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2599360 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3055425 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3055425 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599360 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3132204 | - | - |
