Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133847.3 | NP_598608.2 | 8 - 9 | 1141 | 72 | |
| GenBank mRNA | AB093220.2 | - | 7 - 8 | 885 | 72 | |
| AK047497.1 | - | 8 - 9 | 868 | 72 | ||
| AK048158.1 | - | 8 - 9 | 899 | 72 | ||
| AK083541.1 | - | 8 - 9 | 1141 | 72 | ||
| AK087048.1 | - | 8 - 9 | 894 | 72 | ||
| AK088012.1 | - | 8 - 9 | 897 | 72 | ||
| AK162435.1 | - | 8 - 9 | 902 | 72 | ||
| AK171487.1 | - | 8 - 9 | 899 | 72 | ||
| BC110309.1 | - | 8 - 9 | 897 | 5324218 | 72 | |
| BC116320.2 | - | 8 - 9 | 1093 | 40096002 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tm9sf4
Genomic Map