Assay Details
| Gene Symbol: Bpifb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034875.3 | NP_001030047.2 | 14 - 15 | 1766 | 135 | |
| GenBank mRNA | AK143420.1 | - | 11 - 12 | 1130 | 135 | |
| BC147610.1 | - | 11 - 12 | 1129 | 9056600 | 135 | |
| BC147625.1 | - | 11 - 12 | 1129 | 9056615 | 135 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068615 | AK143420.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2068615 | NM_001034875.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900333 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 369794 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 753572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 164517 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 967113 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 688317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 8072 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96348 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 589899 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814352 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 475667 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 367786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151920 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76015 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068615 | AK143420.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2068615 | NM_001034875.3 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00531172 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04745830 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00531201 | - | - |
| Whole Mouse Genome G4122A | A_52_P164945 | AK143420.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164945 | BC147610.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164945 | BC147625.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P164945 | NM_001034875.3 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757920 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757947 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757949 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757914 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758045 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21758074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21757972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757935 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01238411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01599562 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758025 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757918 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02056078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01262429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757913 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01528062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01812854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757952 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01749808 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757937 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02172320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757977 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01605915 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01599563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757944 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758016 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01749810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758018 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01605921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01892437 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757954 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01749811 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01749813 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757994 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01812860 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02023218 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02023216 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757970 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757941 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757964 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758001 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01892436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757946 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757920 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01700765 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757991 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757929 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757914 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111932 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01749820 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757925 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02172328 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01605917 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757980 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21758030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21757960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477435 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477428 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477433 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477430 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477438 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477429 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477425 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477424 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477431 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477436 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477437 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477427 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477426 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477432 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477434 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477423 | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200018419.mm.1 | NM_001034875.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018419.mm.1 | NM_001034875.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018419.mm.1 | TC0200002330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018416.mm.1 | NM_001034875.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018416.mm.1 | NM_001034875.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018416.mm.1 | TC0200002330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018418.mm.1 | NM_001034875.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018418.mm.1 | NM_001034875.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018418.mm.1 | TC0200002330.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200018417.mm.1 | NM_001034875.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018417.mm.1 | NM_001034875.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200018417.mm.1 | TC0200002330.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 353233 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1250372 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2781850 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2781850 | - | - |

Bpifb4
Genomic Map