| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200018398.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018398.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018398.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018402.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018402.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018402.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018405.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018405.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018405.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018401.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018401.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018401.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018406.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018406.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018406.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018404.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018397.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018397.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018397.mm.1 |
TC0200002330.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200018408.mm.1 |
NM_001034875.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018408.mm.1 |
NM_001034875.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200018408.mm.1 |
TC0200002330.mm.1 |
MTA TC |