Assay Details
| Gene Symbol: Bpifb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034875.3 | NP_001030047.2 | 9 - 10 | 1372 | 78 | |
| GenBank mRNA | AK143420.1 | - | 6 - 7 | 736 | 78 | |
| BC147610.1 | - | 6 - 7 | 735 | 9056600 | 78 | |
| BC147625.1 | - | 6 - 7 | 735 | 9056615 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bpifb4
Genomic Map