Assay Details
| Gene Symbol: Gab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301298.1 | NP_001288227.1 | 8 - 9 | 2687 | 91 | |
| NM_021356.2 | NP_067331.2 | 7 - 8 | 2557 | 91 | ||
| GenBank mRNA | AJ250669.1 | - | 7 - 8 | 1704 | 91 | |
| AK132176.1 | - | 8 - 9 | 2688 | 91 | ||
| AK155423.1 | - | 7 - 8 | 2568 | 91 | ||
| BC007483.1 | - | 7 - 8 | 2557 | 3495926 | 91 | |
| BC094659.1 | - | 7 - 8 | 1908 | 6854020 | 91 | |
| CT010264.1 | - | 7 - 8 | 1681 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gab1
Genomic Map