Assay Details
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027773.3 | NP_082049.1 | 2 - 3 | 283 | 110 | |
GenBank mRNA | AK077374.1 | - | 2 - 3 | 243 | 111 | |
AK160183.1 | - | 2 - 3 | 244 | 110 | ||
BC115519.1 | - | 2 - 3 | 282 | 40101189 | 110 | |
BC115520.1 | - | 2 - 3 | 283 | 40101190 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
