Assay Details
| Gene Symbol: Tlcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291156.1 | NP_001278085.1 | 2 - 3 | 410 | 68 | |
| NM_027249.3 | NP_081525.1 | 2 - 3 | 410 | 68 | ||
| GenBank mRNA | AK008522.1 | - | 2 - 3 | 386 | 68 | |
| BC021945.1 | - | 2 - 3 | 381 | 5098319 | 68 | |
| CV562896.1 | - | 2 - 3 | 411 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tlcd2
Genomic Map