Assay Details
| Gene Symbol: Glod4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026029.3 | NP_080305.2 | 6 - 7 | 687 | 76 | |
| GenBank mRNA | AK006979.1 | - | 6 - 7 | 1010 | 76 | |
| AK007439.1 | - | 6 - 7 | 652 | 76 | ||
| AK012569.1 | - | 6 - 7 | 665 | 76 | ||
| AK013212.1 | - | 6 - 7 | 665 | 76 | ||
| AK138073.1 | - | 664 | 76 | |||
| BC049937.1 | - | 3 - 4 | 316 | 76 | ||
| BC055356.1 | - | 1 - 2 | 60 | 76 | ||
| BC061012.1 | - | 6 - 7 | 679 | 6587019 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Glod4
Genomic Map