Assay Details
| Gene Symbol: Doc2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007873.2 | NP_031899.2 | 4 - 5 | 805 | 69 | |
| GenBank mRNA | BC067030.1 | - | 4 - 5 | 805 | 30534150 | 69 |
| D85037.1 | - | 4 - 5 | 797 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Doc2b
Genomic Map