Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200020203.mm.1 |
NM_172150.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020203.mm.1 |
NM_172150.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020203.mm.1 |
TC0200002477.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020205.mm.1 |
NM_172150.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020205.mm.1 |
NM_172150.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020205.mm.1 |
TC0200002477.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020206.mm.1 |
NM_172150.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020206.mm.1 |
NM_172150.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020206.mm.1 |
TC0200002477.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200020202.mm.1 |
NM_172150.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020202.mm.1 |
NM_172150.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200020202.mm.1 |
TC0200002477.mm.1 |
MTA TC |