Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200019912.mm.1 |
NR_040498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019912.mm.1 |
NR_040498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019912.mm.1 |
TC0200002446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019913.mm.1 |
NR_040498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019913.mm.1 |
NR_040498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019913.mm.1 |
TC0200002446.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200019911.mm.1 |
NR_040498.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019911.mm.1 |
NR_040498.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200019911.mm.1 |
TC0200002446.mm.1 |
MTA TC |