Assay Details
| Gene Symbol: Pltp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011125.2 | NP_035255.1 | 11 - 12 | 1246 | 73 | |
| GenBank mRNA | AK148330.1 | - | 11 - 12 | 1251 | 73 | |
| U28960.1 | - | 10 - 11 | 1104 | 73 | ||
| U37226.1 | - | 11 - 12 | 1176 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pltp
Genomic Map