Assay Details
| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011943.2 | NP_036073.1 | 10 - 11 | 1172 | 61 | |
| GenBank mRNA | AK043482.1 | - | 10 - 11 | 1173 | 61 | |
| AK077712.1 | - | 10 - 11 | 1167 | 61 | ||
| AK140289.1 | - | 11 - 12 | 1157 | 61 | ||
| AK141708.1 | - | 10 - 11 | 1157 | 61 | ||
| AK162671.1 | - | 10 - 11 | 1126 | 61 | ||
| BC075652.1 | - | 10 - 11 | 1171 | 30541969 | 61 | |
| U39066.1 | - | 10 - 11 | 1024 | 61 | ||
| X97052.1 | - | 10 - 11 | 1030 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k6
Genomic Map