Assay Details
| Gene Symbol: Map2k6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011943.2 | NP_036073.1 | 7 - 8 | 828 | 139 | |
| GenBank mRNA | AK043482.1 | - | 7 - 8 | 829 | 139 | |
| AK077712.1 | - | 7 - 8 | 823 | 139 | ||
| AK086722.1 | - | 829 | 139 | |||
| AK138757.1 | - | 7 - 8 | 829 | 139 | ||
| AK140289.1 | - | 8 - 9 | 813 | 139 | ||
| AK141708.1 | - | 7 - 8 | 813 | 139 | ||
| AK162671.1 | - | 7 - 8 | 782 | 139 | ||
| BC075652.1 | - | 7 - 8 | 827 | 30541969 | 139 | |
| U39066.1 | - | 7 - 8 | 680 | 139 | ||
| X97052.1 | - | 7 - 8 | 686 | 139 | ||
| X98067.1 | - | 7 - 8 | 921 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Map2k6
Genomic Map