Assay Details
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030209.3 | NP_084485.1 | 3 - 4 | 571 | 77 | |
GenBank mRNA | AK019034.1 | - | 3 - 4 | 567 | 77 | |
AK033858.1 | - | 3 - 4 | 532 | 77 | ||
AK132171.1 | - | 3 - 4 | 514 | 77 | ||
AK149909.1 | - | 3 - 4 | 528 | 77 | ||
AK149925.1 | - | 3 - 4 | 528 | 77 | ||
AK150180.1 | - | 3 - 4 | 528 | 77 | ||
AK154308.1 | - | 3 - 4 | 571 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
