Assay Details
Gene Symbol: Crispld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030209.3 | NP_084485.1 | 8 - 9 | 1112 | 84 | |
GenBank mRNA | AK019034.1 | - | 8 - 9 | 1108 | 84 | |
AK033858.1 | - | 8 - 9 | 1076 | 84 | ||
AK132171.1 | - | 8 - 9 | 1055 | 84 | ||
AK149909.1 | - | 8 - 9 | 1069 | 84 | ||
AK149925.1 | - | 8 - 9 | 1069 | 84 | ||
AK150180.1 | - | 8 - 9 | 1069 | 84 | ||
AK154308.1 | - | 8 - 9 | 1112 | 84 | ||
BC090642.1 | - | 8 - 9 | 1106 | 30653388 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
