Assay Details
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 34 - 35 | 4574 | 135 | |
GenBank mRNA | AF498362.1 | - | 34 - 35 | 4574 | 135 | |
AK043744.1 | - | 123 | 135 | |||
AK173034.1 | - | 33 - 34 | 4315 | 135 | ||
BC089557.1 | - | 10 - 11 | 1217 | 135 | ||
BC141401.1 | - | 34 - 35 | 4537 | 9056014 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
