Assay Details
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013851.2 | NP_038879.2 | 9 - 10 | 1474 | 110 | |
GenBank mRNA | AF498362.1 | - | 9 - 10 | 1474 | 110 | |
AK138196.1 | - | 9 - 10 | 1475 | 110 | ||
AK173034.1 | - | 8 - 9 | 1215 | 110 | ||
BC141401.1 | - | 9 - 10 | 1437 | 9056014 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
