Assay Details
| Gene Symbol: Git1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004144.1 | NP_001004144.1 | 12 - 13 | 1293 | 58 | |
| GenBank mRNA | BC079870.1 | - | 12 - 13 | 1293 | 6831910 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Git1
Genomic Map