Assay Details
Gene Symbol: Lyzl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027083.1 | NP_081359.1 | 4 - 5 | 565 | 96 | |
GenBank mRNA | AK006279.1 | - | 4 - 5 | 482 | 96 | |
AY566284.1 | - | 3 - 4 | 417 | 96 | ||
BC048617.1 | - | 4 - 5 | 565 | 6742605 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006013 | AK006279.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006013 | NM_027083.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2006013 | BC048617.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 553215 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445914 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928779 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454192 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 255467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 604398 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130154 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137802 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006013 | AK006279.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006013 | NM_027083.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2006013 | BC048617.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504512 | AK006279.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504512 | AY566284.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504512 | NM_027083.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P504512 | BC048617.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P227995 | AK006279.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P227995 | AY566284.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P227995 | NM_027083.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P227995 | BC048617.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086190 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086159 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086151 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086200 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086142 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086173 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086152 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086161 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086164 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086176 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086148 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086153 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086170 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086144 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086178 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086182 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086135 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086155 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086191 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086166 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086183 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086195 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28086168 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07472552 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P137802 | - | - |
Whole Mouse Genome G4122A | A_52_P227995 | AK006279.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P227995 | AY566284.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P227995 | NM_027083.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P227995 | BC048617.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P504512 | AK006279.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P504512 | AY566284.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P504512 | NM_027083.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P504512 | BC048617.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P137802 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188968 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188966 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188979 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188964 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188972 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09188973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086155 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086087 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086069 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086045 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086097 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086084 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086038 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086042 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086173 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086085 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086072 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086165 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086061 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28086094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09288637 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086138 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09355350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086137 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086109 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086134 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086106 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086061 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086072 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09355346 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086063 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09355345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086169 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09188973 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09188978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086059 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086111 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09244843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08988135 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09165193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09244842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09188970 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09739065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086074 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09522781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09688350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09244840 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09696143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P28086113 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09188974 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10391943 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391944 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391947 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391946 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391948 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10391945 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455920_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432482_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1432481_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10391942 | - | - |
Mouse Genome 430 2.0 Array | 1432482_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1455920_x_at | - | - |
Mouse Genome 430 2.0 Array | 1432482_at | - | - |
Mouse Genome 430 2.0 Array | 1455920_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1432481_a_at | - | - |
Mouse Genome 430 2.0 Array | 1432481_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69444_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69444_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1100036269.mm.1 | NM_027083.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100036269.mm.1 | NM_027083.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100036269.mm.1 | TC1100003877.mm.1 | MTA TC |
Mouse Expression Set 430 | 1432481_a_at | - | - |
Mouse Expression Set 430 | 1432481_PM_a_at | - | - |
Mouse Expression Set 430 | 1455920_x_at | - | - |
Mouse Expression Set 430 | 1432482_at | - | - |
Mouse Expression Set 430 | 1432482_PM_at | - | - |
Mouse Expression Set 430 | 1455920_PM_x_at | - | - |
Murine Genome U74v2 Set | 165422_at | - | - |
Murine Genome U74v2 Set | 165422_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432482_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455920_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455920_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432482_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432481_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432481_PM_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 525421 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4480333 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236299 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1236299 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1236299 | - | - |
