Assay Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008952.2 | NP_032978.2 | 3 - 4 | 573 | 56 | |
| GenBank mRNA | AK008555.2 | - | 3 - 4 | 534 | 56 | |
| AK149488.1 | - | 3 - 4 | 522 | 56 | ||
| BC013525.1 | - | 3 - 4 | 487 | 4237443 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pipox
Genomic Map