Assay Details
| Gene Symbol: Pipox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008952.2 | NP_032978.2 | 6 - 7 | 1058 | 83 | |
| GenBank mRNA | AK008555.2 | - | 6 - 7 | 1019 | 83 | |
| AK149488.1 | - | 6 - 7 | 1007 | 83 | ||
| BC013525.1 | - | 6 - 7 | 972 | 4237443 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pipox
Genomic Map