Assay Details
Gene Symbol: Engase | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172573.2 | NP_766161.1 | 8 - 9 | 1145 | 84 | |
GenBank mRNA | AK048667.1 | - | 8 - 9 | 1147 | 84 | |
AK084254.1 | - | 8 - 9 | 1278 | 84 | ||
AK167142.1 | - | 8 - 9 | 1117 | 84 | ||
AK171813.1 | - | 3 - 4 | 469 | 84 | ||
BC060640.1 | - | 7 - 8 | 969 | 84 | ||
BC120923.1 | - | 8 - 9 | 1143 | 84 | ||
BC120924.1 | - | 8 - 9 | 1147 | 40097903 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
