Assay Details
| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022313.2 | NP_071708.2 | 5 - 6 | 623 | 72 | |
| GenBank mRNA | AB049389.1 | - | 5 - 6 | 599 | 72 | |
| AF190731.1 | - | 5 - 6 | 608 | 72 | ||
| AK012155.1 | - | 5 - 6 | 613 | 72 | ||
| BC037603.1 | - | 5 - 6 | 608 | 72 | ||
| BC068271.1 | - | 5 - 6 | 618 | 30008388 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eral1
Genomic Map