Assay Details
| Gene Symbol: Eral1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022313.2 | NP_071708.2 | 6 - 7 | 738 | 73 | |
| GenBank mRNA | AB049389.1 | - | 6 - 7 | 714 | 73 | |
| AK012155.1 | - | 6 - 7 | 728 | 73 | ||
| BC037603.1 | - | 6 - 7 | 723 | 73 | ||
| BC068271.1 | - | 6 - 7 | 733 | 30008388 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Eral1
Genomic Map