Assay Details
| Gene Symbol: Nek8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080849.3 | NP_543125.1 | 4 - 5 | 674 | 75 | |
| GenBank mRNA | AF407579.1 | - | 4 - 5 | 624 | 75 | |
| AK014546.1 | - | 4 - 5 | 635 | 75 | ||
| AK154358.1 | - | 4 - 5 | 637 | 75 | ||
| BC057995.1 | - | 4 - 5 | 619 | 75 | ||
| BC070457.1 | - | 4 - 5 | 623 | 30603080 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nek8
Genomic Map