Assay Details
| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083906.1 | NP_001077375.1 | 5 - 6 | 2382 | 70 | |
| GenBank mRNA | AK136580.1 | - | 2 - 3 | 425 | 70 | |
| BC133713.1 | - | 5 - 6 | 2382 | 40143429 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c2
Genomic Map