Assay Details
| Gene Symbol: Nr3c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083906.1 | NP_001077375.1 | 6 - 7 | 2530 | 109 | |
| GenBank mRNA | AK136580.1 | - | 3 - 4 | 573 | 109 | |
| BC133713.1 | - | 6 - 7 | 2530 | 40143429 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nr3c2
Genomic Map