Assay Details
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028074.4 | NP_082350.3 | 12 - 13 | 1548 | 83 | |
GenBank mRNA | AK049311.1 | - | 12 - 13 | 1550 | 83 | |
AK134182.1 | - | 11 - 12 | 1354 | 83 | ||
AK169816.1 | - | 12 - 13 | 1494 | 83 | ||
AK171730.1 | - | 12 - 13 | 1417 | 83 | ||
BC043036.1 | - | 11 - 12 | 1305 | 5698965 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
