Assay Details
Gene Symbol: Ddx42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028074.4 | NP_082350.3 | 7 - 8 | 970 | 85 | |
GenBank mRNA | AK049311.1 | - | 7 - 8 | 972 | 85 | |
AK134182.1 | - | 6 - 7 | 776 | 85 | ||
AK169816.1 | - | 7 - 8 | 916 | 85 | ||
AK171730.1 | - | 7 - 8 | 839 | 85 | ||
BC043036.1 | - | 6 - 7 | 727 | 5698965 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
