Assay Details
| Gene Symbol: Slc13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054055.2 | NP_473396.1 | 9 - 10 | 1252 | 82 | |
| GenBank mRNA | AK085326.1 | - | 9 - 10 | 1250 | 82 | |
| AK128982.1 | - | 9 - 10 | 1249 | 82 | ||
| AK138344.1 | - | 9 - 10 | 1250 | 82 | ||
| AK157881.1 | - | 9 - 10 | 1221 | 82 | ||
| BC026803.1 | - | 9 - 10 | 1238 | 4238769 | 82 | |
| BC028882.1 | - | 8 - 9 | 1111 | 82 | ||
More Information
Set Membership: |
|

Slc13a3
Genomic Map