Assay Details
| Gene Symbol: Cdh22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174988.3 | NP_778153.2 | 6 - 7 | 1520 | 61 | |
| GenBank mRNA | AB019618.1 | - | 6 - 7 | 1510 | 61 | |
| AK140248.1 | - | 6 - 7 | 1638 | 61 | ||
| BC112407.1 | - | 5 - 6 | 1215 | 40060660 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh22
Genomic Map