Assay Details
| Gene Symbol: Nell1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037906.2 | NP_001032995.1 | 18 - 19 | 2197 | 85 | |
| GenBank mRNA | AK046127.1 | - | 1526 | 85 | ||
| AY622226.1 | - | 18 - 19 | 2197 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nell1
Genomic Map